Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588508 Supplier Invitrogen™ Supplier No. PA588508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAEAAGGKY RSTVSKSKDP SGLLISVIRT LSTSDDVEDR ENEKGRLEEA YEKCDRDLDE LIVQHYTELT TAIRTYQSIT ERITNSRNKI KQVKENLLSC KMLLHCKRDE LRKLWIEGIE HKHVLNLLDE IENIKQVPQK LEQCMASKHY LSATDMLVSA VESLEGPLLQ VEGLSDLRLE LHSKKMNLHL VLIDELHRHL YIKSTSRVVQ RNKEKGKISS LVKDASVPLI DVTNLPTPRK FLDTSHYSTA GSSSVREINL QDIKEDLELD PEENSTLFMG; Positive Samples: A-431, Jurkat, Mouse brain, Mouse thymus, Mouse liver, Rat brain.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EXOC4
Gene Accession No. O35382, Q62824, Q96A65
Gene Alias ALR; augmenter of liver regeneration (ALR) pseudogene; C78892; Exoc4; Exocyst complex component 4; exocyst complex component 4 tv1; Exocyst complex component 4-like protein; exocyst complex component Sec8; KIAA1699; LOW QUALITY PROTEIN: exocyst complex component 4; MGC27170; REC8; rSec8; Sec8; SEC8 protein; Sec8l1; SEC8-like 1; Sec8p; secretory protein SEC8; si:ch211-92l17.2; si:rp71-18a24.2
Gene Symbols EXOC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human EXOC4 (NP_0685793).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116654, 20336, 60412
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.