Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXOC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563904
Description
Immunogen sequence: TVLAKLIQNV FEIKLQSFVK EQLEECRKSD AEQYLKNLYD LYTRTTNLSS KLMEFNLGTD KQTF Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
Specifications
EXOC5 | |
Polyclonal | |
Unconjugated | |
Exoc5 | |
71 kDa component of rsec6/8 secretory complex; AI447711; AI448003; DKFZp666H126; Exoc5; Exocyst complex component 5; exocyst complex component Sec10; Gm76; hSec10; LOC531907 protein; p71; p71 component of rsec6/8 secretory complex; PRO1912; SEC10; SEC10 protein; Sec10l1; SEC10-like 1; SEC10P; zgc:175220 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00471 | |
Exoc5 | |
Recombinant protein corresponding to Human EXOC5. Recombinant protein control fragment (Product #RP-100834). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction