Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109698 Supplier Invitrogen™ Supplier No. PA5109698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIFISYLENY IEVETGYLKS RSAMILQRYY DSKNHQKRSI GTGGIQDLKE RIRQRTNLPL GPSIDTHGET FLSQEVVVNL LQETKQAFER CHRLSDPSDL PRNAFRIFTI LVEFLCIEHI DYALETGLAG IPSSDSRNAN LYFLDVVQQA NTIFHLFDKQ FNDHLMPLIS SSPKLSECLQ KKKEIIEQME MKLDTGIDRT LNCMIGQMKH ILAAEQKKTD FKPEDENNVL IQYTNACVKV CAYVRKQVEK IKNSMDGKNV DTVLMELGVR F.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Exoc5
Gene Accession No. O00471, P97878, Q3TPX4
Gene Alias 71 kDa component of rsec6/8 secretory complex; AI447711; AI448003; DKFZp666H126; Exoc5; Exocyst complex component 5; exocyst complex component Sec10; Gm76; hSec10; LOC531907 protein; p71; p71 component of rsec6/8 secretory complex; PRO1912; SEC10; SEC10 protein; Sec10l1; SEC10-like 1; SEC10P; zgc:175220
Gene Symbols Exoc5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-610 of human EXOC5 (NP_006535.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105504, 10640, 60627
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.