Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544753 Supplier Invitrogen™ Supplier No. PA544753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLEEETDQTY ENVLAEIQSF ELPVEATLRS VYDDQPNAHK KFMEKLDACI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Exoc6
Gene Accession No. Q8R313, Q8TAG9
Gene Alias 4833405E05Rik; AW413330; C430002C19; DKFZp761I2124; Exoc6; EXOC6A; Exocyst complex component 6; exocyst complex component Sec15A; FLJ1125; FLJ11251; hbd; MGC33397; msec15; rSec15; rsec15-like protein; Sec15; SEC15 homolog; SEC15A; SEC15L; SEC15L1; SEC15L3; SEC15-like 1; SEC15-like protein 1; SEC15-like protein 3; Sec15p; zgc:77548
Gene Symbols Exoc6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human EXOC6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107371, 54536
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.