Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557595 Supplier Invitrogen™ Supplier No. PA557595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TFSVSLQKQN KMKFGKNMYI NRDRIPEERN ETVLKHSLEE E Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 56%.

The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Exoc6
Gene Accession No. Q8TAG9
Gene Alias 4833405E05Rik; AW413330; C430002C19; DKFZp761I2124; Exoc6; EXOC6A; Exocyst complex component 6; exocyst complex component Sec15A; FLJ1125; FLJ11251; hbd; MGC33397; msec15; rSec15; rsec15-like protein; Sec15; SEC15 homolog; SEC15A; SEC15L; SEC15L1; SEC15L3; SEC15-like 1; SEC15-like protein 1; SEC15-like protein 3; Sec15p; zgc:77548
Gene Symbols Exoc6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EXOC6. Recombinant protein control fragment (Product #RP-97094).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54536
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.