Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564145 Supplier Invitrogen™ Supplier No. PA564145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNLNQKIDQF LQLADYDWMT GDLGNKASDY LVDLIAFLRS TFAVFTHLPV SGSCYFVLYI Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC6B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOC6B
Gene Accession No. Q9Y2D4
Gene Alias 4930569O18Rik; EXOC6B; Exocyst complex component 6B; exocyst complex component Sec15B; G430127E12Rik; KIAA0919; mKIAA0919; RGD1560638; SEC15 homolog B; Sec15b; Sec15l2; SEC15-like 2; SEC15-like protein 2
Gene Symbols EXOC6B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EXOC6B. Recombinant protein control fragment (Product #RP-101819).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.