Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXOC6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565123
Description
Immunogen sequence: MKQAQQQRNLD NIVLQQPRIG SKRKSKKDAY IIFDTEIEST SPKSEQDSGI LDVEDEE Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.
Specifications
EXOC6B | |
Polyclonal | |
Unconjugated | |
EXOC6B | |
4930569O18Rik; EXOC6B; Exocyst complex component 6B; exocyst complex component Sec15B; G430127E12Rik; KIAA0919; mKIAA0919; RGD1560638; SEC15 homolog B; Sec15b; Sec15l2; SEC15-like 2; SEC15-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2D4 | |
EXOC6B | |
Recombinant Human EXOC6B. Recombinant protein control fragment (Product #RP-104644). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction