Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC6B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9796 Supplier Bioss Supplier No. BS14663R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.

Specifications

Antigen EXOC6B
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene EXOC6B
Gene Accession No. Q9Y2D4
Gene Alias 4930569O18Rik; EXOC6B; Exocyst complex component 6B; Exocyst complex component Sec15B; G430127E12Rik; KIAA0919; mKIAA0919; SEC15 homolog B; Sec15b; Sec15l2; SEC15-like 2; SEC15-like protein 2
Gene Symbols EXOC6B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EXOC6B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23233
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.