Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110882 Supplier Invitrogen™ Supplier No. PA5110882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DHVISYYHVA SDTEKIIREG PTGRLEEYLG SMAKIQKAVE YFQDNSPDSP ELNKVKLLFE RGKEALESEF RSLMTRHSKV VSPVLILDLI SGDDDLEAQE DVTLEHLPES VLQDVIRISR W Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.

The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOC7
Gene Accession No. Q9UPT5
Gene Alias 2-5-3p; EX070; EXO70; Exo70p; EXOC1; Exoc7; Exocyst complex component 7; exocyst complex component Exo70; exocyst component protein 70 kDa homolog; HGNC:23214; KIAA1067; rExo70; sec70; YJL085W
Gene Symbols EXOC7
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EXOC7. Recombinant protein control fragment (Product #RP-93494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23265
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.