Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXOC7 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518508
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.
Specifications
| EXOC7 | |
| Polyclonal | |
| Unconjugated | |
| EXOC7 | |
| 2-5-3p; EX070; EXO70; Exo70p; EXOC1; Exoc7; Exocyst complex component 7; exocyst complex component Exo70; exocyst component protein 70 kDa homolog; HGNC:23214; KIAA1067; rExo70; sec70; YJL085W | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23265 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UPT5 | |
| EXOC7 | |
| Synthetic peptide sequence (DNIKNDPDKEYN) corresponding to the internal amino acids of EXOC7 (aa 481-493). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction