Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114003 Supplier Invitrogen™ Supplier No. PA5114003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMAMSDSGA SRLRRQLESG GFEARLYVKQ LSQQSDGDRD LQEHRQRIQA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Component of the exocyst complex involved in the docking of exocystic vesicles with fusion sites on the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EXOC8
Gene Accession No. Q8IYI6
Gene Alias AI414418; EXO84; Exo84p; Exoc8; Exocyst complex 84 kDa subunit; exocyst complex component 8; R74783; SEC84
Gene Symbols EXOC8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human EXOC8 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149371
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.