Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117941 Supplier Invitrogen™ Supplier No. PA5117941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EXOC8 Polyclonal Antibody for Western Blot, IP

Component of the exocyst complex involved in the docking of exocystic vesicles with fusion sites on the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC8
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene EXOC8
Gene Accession No. Q8IYI6
Gene Alias AI414418; EXO84; Exo84p; Exoc8; Exocyst complex 84 kDa subunit; exocyst complex component 8; R74783; SEC84
Gene Symbols EXOC8
Host Species Rabbit
Immunogen E. coli-derived Human EXOC8 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149371
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.