Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exonuclease 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25829925UL
Description
Exonuclease 1 Polyclonal specifically detects Exonuclease 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Exonuclease 1 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EXOI, exonuclease 1, Exonuclease I, HEX1rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1, hExo1, hExoIEC 3.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
EXO1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDETAVTDKENNLHESEYGDQEGKRLVDTDVARNSSDDIPNNHIPGDHIPDKATVFTDEESYSFKSSKFTRTISPPTLGTLRSCFSWSGGLGDFSRTPSPSPSTALQQFRRKSDSPTSLPENNMSDVSQLK | |
25 μL | |
Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Editing and Processing Endonucleases, Stem Cell Markers, Tumor Suppressors | |
9156 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction