Learn More
EXOSC1 Mouse anti-Human, Clone: OTI4C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808591

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Specifications
EXOSC1 | |
Monoclonal | |
Unconjugated | |
EXOSC1 | |
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51013 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y3B2 | |
EXOSC1 | |
Full length human recombit protein of human EXOSC1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.