Learn More
EXOSC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303885AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Specifications
EXOSC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9DAA6, Q9Y3B2 | |
Exosc1 | |
Between 145 and 195 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
EXOSC1 | |
RP11-452K12.9, CGI-108, CSL4, Csl4p, SKI4, Ski4p, hCsl4p, p13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51013, 66583 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.