Learn More
EXOSC10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | EXOSC10 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6607
![]() |
Bethyl Laboratories, Inc
A303987AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.Specifications
EXOSC10 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4, P100 polymyositis-scleroderma overlap syndrome-associated autoantigen, autoantigen PM-SCL, autoantigen PM/Scl 2, polymyositis/scleroderma autoantigen 100 kDa, polymyositis/scleroderma autoantigen 2 (100kD), polymyositis/scleroderma autoantigen 2, 100kDa | |
Between 200 and 250 | |
Primary | |
4° C | |
EXOSC10 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P56960, Q01780 | |
50912, 5394 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.