Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC2 Mouse anti-Human, Clone: 1G8B1, Proteintech
SDP

Catalog No. 501736535 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6535 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6535 Supplier Proteintech Group Inc Supplier No. 660991IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferase

Specifications

Antigen EXOSC2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G8B1
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOSC2
Gene Accession No. Q13868
Gene Alias EXOSC2, exosome component 2, hRrp4p, p7, RRP4, Rrp4p
Gene Symbols EXOSC2
Host Species Mouse
Immunogen EXOSC2 Fusion Protein Ag7003
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23404
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.