Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC3 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene
SDP

Catalog No. 502540109
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0109 30 μL
50-167-5409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0109 Supplier OriGene Supplier No. TA504795S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EXOSC3 Monoclonal antibody specifically detects EXOSC3 in Human samples. It is validated for Western Blot

Specifications

Antigen EXOSC3
Applications Western Blot
Classification Monoclonal
Clone OTI4C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Exosc3
Gene Accession No. Q9NQT5
Gene Alias 2310005D06Rik; AI593501; bA3J10.7; CGI-102; Exosc3; Exosome complex component RRP40; exosome complex exonuclease RRP40; exosome complex exonuclease RRP40; exosome complex component RRP40; exosome component 3; hRrp-40; hRrp40p; hypothetical protein LOC533245; LOW QUALITY PROTEIN: exosome complex component RRP40; p10; PCH1B; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p
Host Species Mouse
Immunogen Full length human recombinant protein of human EXOSC3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51010
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.