Learn More
EXOSC3 Mouse anti-Human, Clone: OTI4C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504795

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
EXOSC3 is component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA.Specifications
EXOSC3 | |
Monoclonal | |
Unconjugated | |
EXOSC3 | |
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51010 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NQT5 | |
EXOSC3 | |
Full length human recombit protein of human EXOSC3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.