Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588182 Supplier Invitrogen™ Supplier No. PA588182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEPASVAAE SLAGSRARAA RTVLGQVVLP GEELLLPEQE DAEGPGGAVE RPLSLNARAC SRVRVVCGPG LRRCGDRLLV TKCGRLRHKE PGSGSGGGVY WVDSQQKRYV PVKGDHVIGI VTAKSGDIFK VDVGGSEPAS LSYLSFEGAT KRNRPNVQVG DLIYGQFVVA NKDMEPEMVC IDSCGRANGM GVIGQDGLLF KVTLGLIRKL LAPDCEIIQE VGKLYPLEIV FGMNGRIWVK AKTIQQTLIL ANILEACEHM TSDQRKQIFS RLAES; Positive Samples: A-431, HeLa, U-87MG, Raji, SGC-7901, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus, nucleolus.

EXOSC3 is component of the exosome 3'->5' exoribonuclease complex and is required for the 3' processing of the 7S pre-RNA to the mature 5. 8S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Exosc3
Gene Accession No. Q7TQK4, Q9NQT5
Gene Alias 2310005D06Rik; AI593501; bA3J10.7; CGI-102; Exosc3; Exosome complex component RRP40; exosome complex exonuclease RRP40; exosome complex exonuclease RRP40; exosome complex component RRP40; exosome component 3; hRrp-40; hRrp40p; hypothetical protein LOC533245; LOW QUALITY PROTEIN: exosome complex component RRP40; p10; PCH1B; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p
Gene Symbols Exosc3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human EXOSC3 (NP_0571262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313243, 51010, 66362
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.