Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117154 Supplier Invitrogen™ Supplier No. PA5117154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, A-431, Jurkat, LO2 Immunogen sequence: MAGLELLSDQ GYRVDGRRAG ELRKIQARMG VFAQADGSAY IEQGNTKALA VVYGPHEIRG SRARALPDRA LVNCQYSSAT FSTGERKRRP HGDRKSCEMG LQLRQTFEAA ILTQLHPRSQ IDIYVQVLQA DGGTYAACVN AATLAVLDAG IPMRDFVCAC SAGFVDGTAL ADLSHVEEAA GGPQLALALL PASGQIALLE MDARLHEDHL ERVLEAAAQA ARDVHTLLDR VVRQHVREAS ILLGD.

Non-catalytic component of the RNA exosome complex which has 3'->5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding 'pervasive' transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. EXOSC4 binds to ARE-containing RNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EXOSC4
Gene Accession No. Q9NPD3
Gene Alias 1110039I09Rik; 1500001N04Rik; EXOSC4; exosc4 protein; exosome complex component RRP41; exosome complex exonuclease RRP41; exosome complex exonuclease RRP41; exosome complex component RRP41; exosome component 4; exosome component Rrp41; FLJ20591; hRrp41p; p12A; ribosomal RNA processing protein 41; ribosomal RNA-processing protein 41; Rrp41; RRP41A; Rrp41p; SKI6; Ski6p; Unknown (protein for MGC:134035); zgc:73175
Gene Symbols EXOSC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-245 of human EXOSC4 (NP_0619101).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300045, 54512
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.