Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXOSC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5112266

Description
Non-catalytic component of the RNA exosome complex which has 3'->5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding 'pervasive' transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. EXOSC4 binds to ARE-containing RNAs.
Specifications
EXOSC4 | |
Polyclonal | |
Unconjugated | |
EXOSC4 | |
1110039I09Rik; 1500001N04Rik; EXOSC4; exosc4 protein; exosome complex component RRP41; exosome complex exonuclease RRP41; exosome complex exonuclease RRP41; exosome complex component RRP41; exosome component 4; exosome component Rrp41; FLJ20591; hRrp41p; p12A; ribosomal RNA processing protein 41; ribosomal RNA-processing protein 41; Rrp41; RRP41A; Rrp41p; SKI6; Ski6p; Unknown (protein for MGC:134035); zgc:73175 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9NPD3 | |
EXOSC4 | |
E. coli-derived Human EXOSC4 fragment | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction