Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566023
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6023 Supplier Bethyl Laboratories, Inc Supplier No. A303774A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

EXOSC4 belongs to the RNase PH family. It is a component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA. It has a 3′-5′ exonuclease activity.

Specifications

Antigen EXOSC4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene EXOSC4
Gene Accession No. Q9NPD3
Gene Alias exosome complex exonuclease RRP41, Exosome component 4, exosome component Rrp41, FLJ20591, hRrp41p, p12A, ribosomal RNA-processing protein 41, RRP41, RRP41A, Rrp41p, SKI6, Ski6p
Gene Symbols EXOSC4
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54512
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.