Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXOSC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562742
Description
Immunogen sequence: RALFCGVACA LDSDGTLVLD PTSKQEKEAR AVLTFALDSV ERKLLMSSTK GLYSDTELQQ CLAAAQAASQ HVFRFYRESL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Non-catalytic component of the RNA exosome complex which has 3'->5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding 'pervasive' transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. [UniProt].
Specifications
EXOSC5 | |
Polyclonal | |
Unconjugated | |
EXOSC5 | |
Chronic myelogenous leukemia tumor antigen 28; CML28; D7Wsu180e; EXOSC5; Exosome complex component RRP46; exosome complex exonuclease RRP46; exosome complex exonuclease RRP46; exosome complex component RRP46; exosome component 5; exosome component Rrp46; hRrp46p; p12B; Ribosomal RNA-processing protein 46; RRP41B; RRP46; Rrp46p; si:ch211-194n6.2; Unknown (protein for MGC:153707); zgc:153707 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQT4 | |
EXOSC5 | |
Recombinant protein corresponding to Human EXOSC5. Recombinant protein control fragment (Product #RP-98876). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction