Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565967 Supplier Invitrogen™ Supplier No. PA565967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEETHTDAKI RAENGTGSSP RGPGCSLRHF ACEQNLLSRP DGSASFLQGD TSVLAGVYGP AEVKVSKEI Highest antigen sequence identity to the following orthologs - mouse 80%, rat 81%.

Non-catalytic component of the RNA exosome complex which has 3'->5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding 'pervasive' transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOSC5
Gene Accession No. Q9NQT4
Gene Alias Chronic myelogenous leukemia tumor antigen 28; CML28; D7Wsu180e; EXOSC5; Exosome complex component RRP46; exosome complex exonuclease RRP46; exosome complex exonuclease RRP46; exosome complex component RRP46; exosome component 5; exosome component Rrp46; hRrp46p; p12B; Ribosomal RNA-processing protein 46; RRP41B; RRP46; Rrp46p; si:ch211-194n6.2; Unknown (protein for MGC:153707); zgc:153707
Gene Symbols EXOSC5
Host Species Rabbit
Immunogen Recombinant Human EXOSC5. Recombinant protein control fragment (Product #RP-106655).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.