Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6326 Supplier Bethyl Laboratories, Inc Supplier No. A303887A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Component of the exosome 3′->5′ exoribonuclease complex, a complex that degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3′-untranslated regions. Required for the 3′-processing of the 7S pre-RNA to the mature 5.8S rRNA. Has a 3′-5′ exonuclease activity.

Specifications

Antigen EXOSC5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene EXOSC5
Gene Accession No. Q9NQT4
Gene Alias RRP41B, RRP46, Rrp46p, hRrp46p, p12B, chronic myelogenous leukemia tumor antigen 28, exosome complex component RRP46, exosome complex exonuclease RRP46, exosome component Rrp46, ribosomal RNA-processing protein 46
Gene Symbols EXOSC5
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56915
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.