Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983318
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3318 Supplier Bioss Supplier No. BS14668R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen EXOSC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EXOSC5
Gene Accession No. Q9NQT4
Gene Alias Chronic myelogenous leukemia tumor antigen 28; CML28; D7Wsu180e; Exosc5; Exosome complex component RRP46; exosome complex exonuclease RRP46; Exosome component 5; exosome component 5-like; exosome component Rrp46; hRrp46p; LOC100357530; p12B; Ribosomal RNA-processing protein 46; RRP41B; RRP46; Rrp46p; si:ch211-194n6.2; Unknown (protein for MGC:153707); zgc:153707
Gene Symbols EXOSC5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EXOSC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.