Learn More
Invitrogen™ EXOSC8 Monoclonal Antibody (4B3)

Description
Peptide Sequence: AEVNLKKKSY LNIRTHPVAT SFAVFDDTLL IVDPTGEEEH LATGTLTIVM DEEGKLCCLH KPGGSGLTGA KLQDCMSRAV TRHKEVKKLM DEVIKSMKPK.
Specifications
Specifications
| Antigen | EXOSC8 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4B3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EXOSC8 |
| Gene Accession No. | Q96B26 |
| Gene Alias | 2310032N20Rik; bA421P11.3; CBP-interacting protein 3; CIP3; EAP2; EXOSC8; Exosome complex component RRP43; exosome complex exonuclease RRP43; exosome component 8; mKIAA4013; OIP2; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; PCH1C; Ribosomal RNA-processing protein 43; RP11-421P11.3; RRP43; Rrp43p |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.