Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXOSC9 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | EXOSC9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6331
![]() |
Bethyl Laboratories, Inc
A303888AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a component of the human exosome, a exoribonuclease complex which processes and degrades RNA in the nucleus and cytoplasm. This component may play a role in mRNA degradation and the polymyositis/scleroderma autoantigen complex. Alternative splicing results in multiple transcript variants.Specifications
EXOSC9 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
EXOSC9; exosome component 9; PM/Scl-75; PMSCL1; RRP45; Rrp45p; p5; p6; P75 polymyositis-scleroderma overlap syndrome associated autoantigen; PMSCL autoantigen, 75kD; autoantigen PM/Scl 1; polymyositis/scleroderma autoantigen 1 (75kD); polymyositis/scleroderma autoantigen 1, 75kDa | |
Between 389 and 439 | |
Primary | |
4° C | |
EXOSC9 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q06265, Q9JHI7 | |
50911, 5393 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title