Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222239PCP
Description
Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Exosome component 1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
The immunogen this antibody was made to, maps to a region between residue 145 and 195 of human Exosome Component 1 using the numbering given in entry NP_057130.1 (GeneID 51013). | |
0.1 mL | |
Immunology | |
51013 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction