Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 10 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297516APC
Description
Exosome component 10 Polyclonal antibody specifically detects Exosome component 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Exosome component 10 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
autoantigen PM-SCL, EC 3.1.13.-, exosome component 10, P100 polymyositis-scleroderma overlap syndrome-associated autoantigen, p2, p3, p4, PM/Scl-100Autoantigen PM/Scl 2, PM-Scl, PMSCL2PMSCL, Polymyositis/scleroderma autoantigen 2, polymyositis/scleroderma autoantigen 2 (100kD), polymyositis/scleroderma autoantigen 2, 100kDa, RRP6, Rrp6p, RRP6Polymyositis/scleroderma autoantigen 100 kDa | |
Produced in rabbits immunized with E. coli-derived Human Exosome component 10 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
5394 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction