Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 5 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222241DL594
Description
Exosome component 5 Polyclonal antibody specifically detects Exosome component 5 in Human samples. It is validated for Western BlotSpecifications
Exosome component 5 | |
Polyclonal | |
Western Blot | |
CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46 | |
The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Exosome Component 5 using the numbering given in entry NP_064543.3 (GeneID 56915). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing, Epigenetics | |
56915 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction