Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 5 Antibody (V80P1C10*C), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250173AF488
Description
Exosome component 5 Monoclonal specifically detects Exosome component 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
Exosome component 5 | |
Monoclonal | |
Alexa Fluor 488 | |
50 mM Sodium Borate with 0.05% Sodium Azide | |
EXOSC5 | |
Peptide Sequence - KLLMSSTKGLY | |
0.1 mL | |
DNA replication Transcription Translation and Splicing, Epigenetics | |
56915 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
V80P1C10*C | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction