Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 7 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297525MFV450
Description
Exosome component 7 Polyclonal antibody specifically detects Exosome component 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Exosome component 7 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
Produced in rabbits immunized with E. coli-derived Human Exosome component 7 fragment. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
23016 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction