Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 7 Antibody (OTI1E8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271720 Supplier Novus Biologicals Supplier No. NBP271720
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Exosome component 7 Monoclonal specifically detects Exosome component 7 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Exosome component 7
Applications Western Blot
Classification Monoclonal
Clone OTI1E8
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:4000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42
Gene Symbols EXOSC7
Host Species Mouse
Immunogen Full length human recombinant protein of human EXOSC7(NP_055819) produced in HEK293T cell.
Molecular Weight of Antigen 31.6 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23016
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.