Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 7 Antibody (OTI1E8), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271720PCP
Description
Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Exosome component 7 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI1E8 | |
Western Blot | |
EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
Full length human recombinant protein of human EXOSC7(NP_055819) produced in HEK293T cell. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
23016 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction