Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Exosome Standards (LnCAP cell line)

Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Specifications
Specifications
Content And Storage | Store at 4°C. After reconstitution store at -70°C. |
Concentration | LYOPH |
Quantity | 6 x 100 μg Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction