Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Exosome Standards (SK-N-SH cell line)
SDP

Catalog No. NB398337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398337 6 x 100 μg Vials
NB398339 6 x 30 μg Vials
NB398336 2 x 30 μg Vials
NB398338 2 x 100 μg Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398337 Supplier Novus Biologicals™ Supplier No. NBP249852600UG
Add to Cart
Add to Cart

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 6 x 100 μg Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.