Learn More
Restek™ EXP Direct Connect Holder
SDP
Catalog No. 06611938 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
EXP Guard Cartridges
Catalog No. For Use With (Equipment)
06-611-938 EXP Guard Cartridges
1 options
Catalog No. 06-611-938 Supplier Restek™ Supplier No. 25808
Free-Turn architecture allows for changing cartridges by hand without breaking inlet/outlet fluid connections
  • Patented titanium hybrid ferrules can be installed repeatedly without compromising high-pressure seal
  • Auto-adjusting design provides ZDV (zero dead volume) connection to any 10-32 female port
  • EXP direct connect holder requires separate guard column cartridges
    • Available from Restek in 2.1, 3, and 4.6 mm
  • Pair with EXP hand-tight fitting for tool-free installation
Specifications
Type EXP Direct Connect Holder
Includes Hex-head fitting and two ferrules
For Use With (Equipment) EXP Guard Cartridges