Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ EXPERTTM Standard Pipette Tips - Sterilized MegaPack
GREENER_CHOICE

Catalog No. F1732071G
Click to view available options
Graduations:
10, 50, 100μL
2, 5, 10μL
Length (Metric):
4.534cm
5.144cm
Model:
E200ST
EL10ST
This item is not returnable. View return policy
This item is not returnable. View return policy

Feature patented, low-retention polymer that creates hydrophobic surface to minimize sample binding while dispensing

  • For high-throughput multichannel pipetting
  • Features 192 tips per layer, 5 layers high
  • Gives choice to expose only 96 tips at a time w/portioned cover, or remove cover completely to access 192 tips
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens

Specifications

Description Sterilized MegaPack
Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model EL10ST
Length (Metric) 4.534cm
Clean Claims RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
For Use With (Application) Pipetting
Compatibility Universal Fit
Format Rack
Tip Style Standard
Packaging Racked
Quantity 960/Cs.
Product Type Air Displacement Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line EXPERT
Material Polypropylene
Shape Conical
Graduations 2, 5, 10μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.