Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exploring with a Microscope CD-ROM
SCI_ED

Catalog No. S32015
Click to view available options
:
Exploring W/microscp Cd Indivi

Conduct actual experiments in your own cyber lab!

  • Interactive CD-ROM
  • Perform a variety of lab simulations on the computer
  • Perform experiments in a virtual lab environment and learn microtechniques including: measurement, use of a counting cell, wet mount preparation, permanent wholemount preparation without the use of hazardous chemicals, hanging drop preparation, Rheinberg illumination, darkfield illumination, and polarization
  • Presents information on the workings of a microscope
  • Includes text, illustrations, video clips, and animation to familiarize the student with basic microtechnique concepts

Exploring with a Microscope Software Combination
Bring your classroom into interactive education by combining all of the hands-on attributes of the Exploring with a Microscope laboratory investigation kit, with its corresponding Neo/Lab™ CD-ROM. The combination set couples all of the benefits of each format - at a discount from the combined individual prices!

Specifications

Grade 6 to 12
Certifications/Compliance NSES
Compatibility Windows™ 98 or greater, Mac™ 8.6 or higher (classic mode)
Category Earth Science
Format CD-ROM
For Use With (Application) Provides students with the ability to use a virtual microscope and perform a variety of lab simulations on the computer
Includes Text, illustrations, video clips, and animation to familiarize the student with basic microtechnique concepts
License Single User
Memory 64MB
Product Type Exploring with a microscope
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.