Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Exploring the Genetics of Taste: SNP Analysis of the PTC Gene Using PCR Experiment Kit
SDPSCI_ED

Catalog No. s29018 Shop All EDVOTEK Products
Click to view available options
Class Size:
25 Students

Catalog No. S29018

Supplier: Edvotek™ 345

Only null left
Add to Cart
Add to Cart

Includes

Instructions, PCR EdvoBeads™, Universal DNA Buffer, TE Buffer, LyphoPrimer™ Mix, LyphoTemplate™ Control DNA, Dryzymes™ Restriction Enzyme HaeIII, Restriction Enzyme Dilution Buffer, 100 bp Ladder, Proteinase K, PTC Paper, Control Taste Paper, UltraSpec-Agarose, Electrophoresis Powder Buffer, InstaStain™ Ethidium Bromide, Disposable plastic cups, 15 ml Conical Tube, Snap-top microcentrifuge tubes, microcentrifuge tubes (1.5 ml screw-cap), 0.2 ml PCR tubes, wax beads, and salt packets

Requires

Thermal cycler, electrophoresis apparatus, power supply, automatic micropipet, microwave or hot plate, waterbath, UV transilluminator.

To identify the presence of the single nucleotide polymorphism (SNP) in an amplified segment of the PTC gene that links detection of the characteristic taste of PTC paper. Edvotek™ Exploring the Genetics of Taste: SNP Analysis of the PTC Gene Using PCR Experiment Kit includes 5 modules.

Five Modules Include:

  • Extraction of DNA from buccal cells
  • Amplifying the segment that contains the polymorphic nucleotide
  • Digestion of the amplified fragment with the restriction enzyme that recognizes the SNP
  • Analysis by gel electrophoresis
  • Tasting the PTC paper to confirm the results obtained
  • For 25 reactions
  • Complete in 3 hrs. 30 min.
  • Level: Advanced
  • Requires wet ice shipment for next day delivery

Specifications

Grade 9 to Undergraduate
Includes Instructions, primer mix, DNA template, PCR EdvoBead™, DNA size ladder, Restriction Enzyme, Restriction Enzyme Reaction and Dilution Buffers, Lysis buffer, agarose, electrophoresis buffer, InstaStain™ Ethidium Bromide, PTC paper.
Quantity 25 Reactions
Time Range Complete in 3 hrs. 30 min.
Content And Storage Freezer Storage (Some components)
Class Size 25 Students
Product Type Exploring the Genetics of Taste: SNP Analysis of the PTC Gene Using PCR Experiment Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.