Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exportin 4 Antibody, Novus Biologicals™
SDP

Catalog No. p-5069270 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056495T 0.025 mg
NB10056495 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056495T Supplier Novus Biologicals Supplier No. NB10056495SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Exportin 4 Polyclonal specifically detects Exportin 4 in Human, Mouse, Rat, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Exportin 4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q9C0E2
Gene Alias exp4, exportin 4, exportin-4, KIAA1721FLJ13046
Gene Symbols XPO4
Host Species Rabbit
Immunogen A portion of amino acids 850-950 of human Exportin-4 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64328
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c), rhesus monkey, chimp (isoforms 1-6), chicken, dog, mouse and rat, 94% homologous in bovine and xenopus, and 88% homologous in zebrafish.
Target Species Human, Mouse, Rat, Canine, Chicken, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.