Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exportin-5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312943
This item is not returnable.
View return policy
Description
Exportin-5 Polyclonal antibody specifically detects Exportin-5 in Human, Amphibian samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Exportin-5 | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Amphibian | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Exp5, exportin 5, exportin-5, FLJ14239, FLJ32057, FLJ45606, KIAA1291exp5, Ran-binding protein 21, RANBP21 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Exportin-5. The exact sequence is proprietary. | |
| 100 μg | |
| Epigenetics | |
| 57510 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction