Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exportin-5 Rat anti-Human, Clone: EX5 4E1, MilliporeSigma™

Catalog No. MABE192225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE192225 25 μL
MABE1922100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE192225 Supplier MilliporeSigma Supplier No. MABE192225UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Exportin-5 Monoclonal antibody specifically detects Exportin-5 in Human samples. It is validated for ELISA, Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exportin-5
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone EX5 4E1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human Exportin-5.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.