Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXPRESS qPCR Supermix, universal

Catalog No. 11785200
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
200 Reactions
2500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-785-200 200 Reactions
11-785-01K 2500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-785-200 Supplier Invitrogen™ Supplier No. 11785200
  • $1,020.00
    $989.65 / Each of 1

    Save $30.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays.

  • EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays
  • This kit comes with a separate tube of ROX reference dye
  • EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available

This kit provides

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, Eppendorf™ MasterCycler™ RealPlex, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Product Type Express qPCR Supermix
Quantity 200 Reactions
Sufficient For 200 Reactions
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less
I used the EXPRESS qPCR Supermix, universal and the PCR efficiency was below 90%. What is the problem?

This indicates that the PCR conditions were suboptimal. Please ensure that the reagents you are using have not been freeze-thawed multiple times and have not remained at room temperature for too long. Verify that the amount of primers you are using is correct.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.