Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXPRESS SYBR™ GreenER™ qPCR Supermix, universal

Catalog No. 1178401K
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-784-01K 1000 Reactions
11-784-200 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-784-01K Supplier Invitrogen™ Supplier No. 1178401K
Only null left
Add to Cart
Add to Cart

Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.

EXPRESS SYBR GreenER qPCR SuperMix Universal comes with separate tubes of ROX reference dye.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains 5 × 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix Universal (2X) and 5 × 500 μl ROX Reference Dye. Sufficient reagents are provided for 2500 reactions of 20 μL each.

Store all components at -20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Product Line SYBR GreenER
Product Type Real Time PCR SYBR Master Mix
Quantity 1,000 reactions
Sufficient For 2500 Reactions of 20 μL Each
For Use With (Application) Gene Expression
No. of Reactions 1000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 5 x 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.