Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527574 Supplier Invitrogen™ Supplier No. PA527574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, Jurkat, NT2D1. Predicted reactivity: Mouse (98%), Rat (96%), Zebrafish (84%), Xenopus laevis (88%), Chicken (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one of two glycosyltransferases involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type II form of multiple exostoses. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXT2
Applications Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene EXT2
Gene Accession No. Q93063
Gene Alias acetylglucosaminyltransferase; AI893565; exostoses (multiple) 2; exostosin 2; exostosin glycosyltransferase 2; exostosin-2; EXT2; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; multiple exostoses protein 2; Multiple exostoses protein 2 homolog; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; putative tumor suppressor protein EXT2; SOTV; SSMS
Gene Symbols EXT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EXT2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.