Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Extended Capillary Column Nuts
SDP

Catalog No. 06611217 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Capillary Column Nut
Extended Capillary Column Nut Kit
Extended Reducing Nut
For Use With (Equipment):
Compact Ferrules
Standard Ferrules
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
06-611-217 Extended Reducing Nut -
06-611-219 Capillary Column Nut Compact Ferrules
06-611-216 Extended Capillary Column Nut Kit Compact Ferrules
06-611-218 Capillary Column Nut Standard Ferrules
06-611-215 Extended Capillary Column Nut Kit Standard Ferrules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-611-217 Supplier Restek™ Supplier No. 22635
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Visible below inlet insulating cup for easy, breakage-free column installation without having to remove the cup

  • Finger-tight design for wrench-free use
  • Available for Standard (1/16 in.) and Compact (Agilent-style) ferrules
  • Made from high-quality stainless steel
  • Meets or exceeds original manufacturer's performance

Specifications

Material Stainless Steel
Type Extended Reducing Nut
Color Silver
Description Finger-Tight Design
For Use With (Application) Agilent GCs
Includes Extended Reducing Nut
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.