Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXTL1 Monoclonal Antibody (353713)
GREENER_CHOICE

Catalog No. PIMA524098
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524098 Supplier Invitrogen™ Supplier No. MA524098
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human (rh) EXTL1, rhEXTL2, rhEXTL3, or recombinant mouse EXTL2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene is a member of the multiple exostoses (EXT) family of glycosyltransferases, which function in the chain polymerization of heparan sulfate and heparin. The encoded protein harbors alpha 1,4- N-acetylglucosaminyltransferase activity, and is involved in chain elongation of heparan sulfate and possibly heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXTL1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 353713
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Extl1
Gene Accession No. Q9JKV7
Gene Alias alpha-N-acetylglucosaminyltransferase II; D430033M16Rik; EC 2.4.1.224; exostoses (multiple)-like 1; exostoses-like 1; exostosin like glycosyltransferase 1; exostosin-L; Exostosin-like 1; Exostosin-like 1 (Exostosin-L) (Multiple exostosis-like protein); exostosin-like glycosyltransferase 1; EXTL; Extl1; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase; glucuronyl-N-acetylglucosaminylproteoglycan alpha-1,4-N- acetylglucosaminyltransferase; MGC70794; multiple exostoses-like 1; multiple exostosis-like protein
Gene Symbols Extl1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 1/EXTL1 Ala30-Pro669.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56219
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.